Tisk…

o-Tolyl isothiocyanate 98%

Dodavatel: Apollo Scientific
Nebezpečí

OR28816-25G OR28816-5G
APOSOR28816-25GEA 5200 CZK
APOSOR28816-25G APOSOR28816-5G
o-Tolyl isothiocyanate 98%
o-Tolyl isothiocyanate
Vzorec: C₈H₇NS
Bod vzplanutí: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00004802
CAS číslo: 614-69-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR