Tisk…

2-Methyl-1H-imidazole-4-carbaldehyde

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Methyl-1H-imidazole-5-carbaldehyde

OR28795-5G
APOSOR28795-5GEA 8900 CZK
APOSOR28795-5G
2-Methyl-1H-imidazole-4-carbaldehyde
2-Methyl-1H-imidazole-4-carbaldehyde
Vzorec: C₅H₆N₂O
MW: 110,12 g/mol
Bod tání: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00173725
CAS číslo: 35034-22-1
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR