1,4-Bis(4-methyl-5-phenyloxazol-2-yl)benzene
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Dimethyl-POPOP, 1,4-Bis[2-(4-methyl-5-phenyloxazolyl)]benzene
OR28759-25G
OR28759-5G
APOSOR28759-25GEA
24800
CZK
APOSOR28759-25G
APOSOR28759-5G
1,4-Bis(4-methyl-5-phenyloxazol-2-yl)benzene
1,4-Bis(4-methyl-5-phenyloxazol-2-yl)benzene
Vzorec:
C₂₆H₂₀N₂O₂ MW: 392.46 Bod tání: 230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005310 CAS číslo: 3073-87-8 EINECS: 221-349-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...