2,2'-Bipyridyl 99+%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR28749-100G
OR28749-250G
OR28749-25G
APOSOR28749-100GEA
1240
CZK
APOSOR28749-100G
APOSOR28749-250G
APOSOR28749-25G
2,2'-Bipyridyl 99+%
2,2'-Bipyridyl
Vzorec:
C₁₀H₈N₂ Bod varu: 273 °C Bod tání: 70…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006212 CAS číslo: 366-18-7 EINECS: 206-674-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...