Ethyl-6-bromohexanoate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Bromocaproic acid ethyl ester, 6-Bromohexanoic acid ethyl ester, Ethyl 6-bromohexanoate
OR28740-100G
OR28740-500G
APOSOR28740-100GEA
1950
CZK
APOSOR28740-100G
APOSOR28740-500G
Ethyl-6-bromohexanoate
Ethyl-6-bromohexanoate
Vzorec:
BrH₂C(CH₂)₄COOCH₂CH₃ MW: 223,11 g/mol Bod varu: 128…130 °C/16mmHg Hustota: 1,254 g/cm³ (25 °C) Bod vzplanutí: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000270 CAS číslo: 25542-62-5 EINECS: 247-085-2 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...