5-Methylpyrazine-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR28728-100G
OR28728-25G
OR28728-5G
APOSOR28728-100GEA
1120
CZK
APOSOR28728-100G
APOSOR28728-25G
APOSOR28728-5G
5-Methylpyrazine-2-carboxylic acid
5-Methylpyrazine-2-carboxylic acid
Vzorec:
C₆H₆N₂O₂ MW: 138,13 g/mol Bod tání: 165…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068241 CAS číslo: 5521-55-1 EINECS: 413-260-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...