Methyl-2-(chlorosulphonyl)benzoate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Methyl 2-(chlorosulphonyl)benzoate
OR28691-1G
OR28691-5G
APOSOR28691-1GEA
622
CZK
APOSOR28691-1G
APOSOR28691-5G
Methyl-2-(chlorosulphonyl)benzoate
Methyl-2-(chlorosulphonyl)benzoate
Vzorec:
C₈H₇ClO₄S Bod tání: 59…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009797 CAS číslo: 26638-43-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...