4-Chloro-2-cyclohexylphenol
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR28650-10G
OR28650-1G
OR28650-25G
OR28650-100G
OR28650-500G
APOSOR28650-10GEA
2240
CZK
APOSOR28650-10G
APOSOR28650-1G
APOSOR28650-25G
APOSOR28650-100G
APOSOR28650-500G
4-Chloro-2-cyclohexylphenol
4-Chloro-2-cyclohexylphenol
Vzorec:
C₁₂H₁₅ClO Bod varu: 170…180 °C/18mmHg Bod tání: 59…60 °C Storage Temperature: Ambient |
CAS číslo:
13081-17-9 EINECS: 235-988-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...