α,3-Dichlorotoluene 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-Chloro-3-(chloromethyl)benzene, 3-Chlorobenzyl chloride, 3-Chlorobenzylchloride
OR28625-100G
OR28625-500G
APOSOR28625-100GEA
992
CZK
APOSOR28625-100G
APOSOR28625-500G
α,3-Dichlorotoluene 98%
α,3-Dichlorotoluene
Vzorec:
C₇H₆Cl₂ MW: 161,03 g/mol Bod varu: 214…216 °C Hustota: 1,27 g/cm³ (25 °C) Bod vzplanutí: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000905 CAS číslo: 620-20-2 EINECS: 210-629-4 UN: 2235 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...