Ethyl butyrylacetate 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl Butyrylacetate, Ethyl 3-oxohexanoate
OR28623-25G
OR28623-100G
OR28623-500G
APOSOR28623-25GEA
622
CZK
APOSOR28623-25G
APOSOR28623-100G
APOSOR28623-500G
Ethyl butyrylacetate 95%
Ethyl butyrylacetate
Vzorec:
C₈H₁₄O₃ Bod varu: 94…95 °C/12mmHg Bod tání: –44 °C Hustota: 0,99 g/cm³ (25 °C) Bod vzplanutí: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009401 CAS číslo: 3249-68-1 EINECS: 221-835-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...