2-(2-Aminoethyl)pyridine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Pyridylethylamine, 2-(2-Pyridyl)ethylamine, 2-Pyridin-2-ylethanamine
OR28599-100G
OR28599-25G
APOSOR28599-100GEA
4860
CZK
APOSOR28599-100G
APOSOR28599-25G
2-(2-Aminoethyl)pyridine 98%
2-(2-Aminoethyl)pyridine
Vzorec:
C₇H₁₀N₂ MW: 122,17 g/mol Bod varu: 208 °C Hustota: 1,021 g/cm³ (25 °C) Bod vzplanutí: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006367 CAS číslo: 2706-56-1 EINECS: 220-295-1 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...