Tisk…

5-Bromoisatin

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Bromoindoline-2,3-dione, 5-Bromo-1H-indole-2,3-dione

OR28576-1G
APOSOR28576-1GEA 622 CZK
APOSOR28576-1G
5-Bromoisatin
5-Bromoisatin
Vzorec: C₈H₄BrNO₂
MW: 226,03 g/mol
Bod tání: 247…253 °C
Storage Temperature: Ambient
MDL Number: MFCD00149345
CAS číslo: 87-48-9
EINECS: 201-747-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR