4-Amino-3-hydroxybenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR2857-25G
OR2857-100G
OR2857-5G
APOSOR2857-25GEA
1330
CZK
APOSOR2857-25G
APOSOR2857-100G
APOSOR2857-5G
4-Amino-3-hydroxybenzoic acid
4-Amino-3-hydroxybenzoic acid
Vzorec:
C₇H₇NO₃ MW: 153.14 Bod tání: 211…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017094 CAS číslo: 2374-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...