Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl 1-Benzyl-3-oxo-4-piperidinecarboxylate HCl, Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, Ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride
OR2854-100G
OR2854-25G
OR2854-5G
APOSOR2854-100GEA
3740
CZK
APOSOR2854-100G
APOSOR2854-25G
APOSOR2854-5G
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride 98%
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride
Vzorec:
C₁₅H₂₀ClNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00012792 CAS číslo: 52763-21-0 EINECS: 258-162-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...