Tisk…

2-Acetylbenzofuran

Dodavatel: Apollo Scientific

OR28532-100G OR28532-25G
APOSOR28532-100GEA 10250 CZK
APOSOR28532-100G APOSOR28532-25G
2-Acetylbenzofuran
2-Acetylbenzofuran
Vzorec: C₁₀H₈O₂
Bod varu: 110…113 °C/3mmHg
Bod tání: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00005849
CAS číslo: 1646-26-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR