Tisk…

Phenylmaleic anhydride

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Phenylfuran-2,5-dione, 3-Phenyl-2,5-furandione

OR28465-1G OR28465-25G OR28465-5G
APOSOR28465-1GEA 622 CZK
APOSOR28465-1G APOSOR28465-25G APOSOR28465-5G
Phenylmaleic anhydride
Phenylmaleic anhydride
Vzorec: C₁₀H₆O₃
MW: 174,16 g/mol
Bod varu: 122…125 °C/0,2mmHg
Bod tání: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00015472
CAS číslo: 36122-35-7
EINECS: 252-881-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR