6-Chloro-7-deazapurine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR2836-25G
OR2836-5G
OR2836-100G
APOSOR2836-25GEA
827
CZK
APOSOR2836-25G
APOSOR2836-5G
APOSOR2836-100G
6-Chloro-7-deazapurine
6-Chloro-7-deazapurine
Vzorec:
C₆H₄ClN₃ Bod tání: 188…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD01686865 CAS číslo: 3680-69-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...