Tisk…

Cyclohexanesulphonyl chloride 90%

Dodavatel: Apollo Scientific
Nebezpečí

OR2831-1G OR2831-25G OR2831-5G
APOSOR2831-1GEA 662 CZK
APOSOR2831-1G APOSOR2831-25G APOSOR2831-5G
Cyclohexanesulphonyl chloride 90%
Cyclohexanesulphonyl chloride
Vzorec: C₆H₁₁ClO₂S
Hustota: 1,259 g/cm³ (25 °C)
Bod vzplanutí: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04117775
CAS číslo: 4837-38-1
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR