4-Mercaptobenzoic acid 90%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Thiobenzoic acid
OR28166-25G
OR28166-5G
OR28166-10G
APOSOR28166-25GEA
6850
CZK
APOSOR28166-25G
APOSOR28166-5G
APOSOR28166-10G
4-Mercaptobenzoic acid 90%
4-Mercaptobenzoic acid
Vzorec:
C₇H₆O₂S MW: 154,1863 g/mol Bod tání: 215…224 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00016617 CAS číslo: 1074-36-8 UN: 3335 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...