4-Amino-3-bromopyridine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Bromo-4-pyridylamine, 3-Bromopyridin-4-amine
OR2801-1G
OR2801-250MG
APOSOR2801-1GEA
827
CZK
APOSOR2801-1G
APOSOR2801-250MG
4-Amino-3-bromopyridine
4-Amino-3-bromopyridine
Vzorec:
C₅H₅BrN₂ MW: 173,01 g/mol Bod tání: 61…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD02068297 CAS číslo: 13534-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...