Tisk…

5-Chloro-2-hydroxybenzophenone

Dodavatel: Apollo Scientific

OR27959-100G OR27959-25G
APOSOR27959-100GEA 11250 CZK
APOSOR27959-100G APOSOR27959-25G
5-Chloro-2-hydroxybenzophenone
5-Chloro-2-hydroxybenzophenone
Vzorec: C₁₃H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00020134
CAS číslo: 85-19-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR