Tisk…

2-Coumaranone 98%

Dodavatel: Apollo Scientific

Synonyms: Benzofuran-2(3H)-one, Benzo[b]furan-2(3H)-one

OR27929-100G OR27929-500G OR27929-1KG
APOSOR27929-100GEA 622 CZK
APOSOR27929-100G APOSOR27929-500G APOSOR27929-1KG
2-Coumaranone 98%
2-Coumaranone
Vzorec: C₈H₆O₂
MW: 134,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005856
CAS číslo: 553-86-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR