3-Methylbenzofuran
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR27821-1G
OR27821-5G
APOSOR27821-1GEA
1660
CZK
APOSOR27821-1G
APOSOR27821-5G
3-Methylbenzofuran
3-Methylbenzofuran
Vzorec:
C₉H₈O MW: 132,16 g/mol Bod varu: 195…197 °C Hustota: 1,046 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00205641 CAS číslo: 21535-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...