Tisk…

3-Aminopicolinic acid 97%, Technical Grade

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Amino-2-pyridinecarboxylic acid, 3-Aminopyridine-2-carboxylic acid, 2-Pyridinecarboxylic acid, 3-amino-

OR2780T-5G OR2780T-25G
APOSOR2780T-5GEA 992 CZK
APOSOR2780T-5G APOSOR2780T-25G
3-Aminopicolinic acid 97%, Technical Grade
3-Aminopicolinic acid
Vzorec: C₆H₆N₂O₂
Bod tání: 218…222 °C
Storage Temperature: Freezer
MDL Number: MFCD00090153
CAS číslo: 1462-86-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR