Tisk…

Benzylammonium chloride 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Benzylamine hydrochloride

OR27725-25G OR27725-100G
APOSOR27725-25GEA 662 CZK
APOSOR27725-25G APOSOR27725-100G
Benzylammonium chloride 99%
Benzylammonium chloride
Vzorec: C₇H₁₀ClN
MW: 143.62
Bod tání: 262…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00012852
CAS číslo: 3287-99-8
EINECS: 221-943-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR