Metacetamol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
N-(3-Hydroxyphenyl)acetamide, N-Acetyl-3-aminophenol, Paracetamol, 3-Acetamidophenol, 3'-Hydroxyacetanilide
OR27724-100G
OR27724-25G
OR27724-5G
APOSOR27724-100GEA
3310
CZK
APOSOR27724-100G
APOSOR27724-25G
APOSOR27724-5G
Metacetamol
Metacetamol
Vzorec:
C₈H₉NO₂ MW: 151,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002263 CAS číslo: 621-42-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...