Tisk…

7-Methoxynaphthalen-1-ol 95%

Dodavatel: Apollo Scientific
Varování

OR27684-1G OR27684-5G
APOSOR27684-1GEA 2280 CZK
APOSOR27684-1G APOSOR27684-5G
7-Methoxynaphthalen-1-ol 95%
7-Methoxynaphthalen-1-ol
Vzorec: C₁₁H₁₀O₂
MW: 174,200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01314215
CAS číslo: 67247-13-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR