Tisk…

2-(Dimethylamino)-5-pyrimidineboronic acid pinacol ester

Dodavatel: Apollo Scientific

Synonyms: N,N-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

OR2768-1G OR2768-5G OR2768-25G
APOSOR2768-1GEA 3310 CZK
APOSOR2768-1G APOSOR2768-5G APOSOR2768-25G
2-(Dimethylamino)-5-pyrimidineboronic acid pinacol ester
2-(Dimethylamino)-5-pyrimidineboronic acid pinacol ester
Vzorec: C₁₂H₂₀BN₃O₂
MW: 249,12 g/mol
Bod tání: 107…112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368238
CAS číslo: 1032759-30-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR