3,4-Dichloro-2,6-dinitrophenol
Dodavatel: Apollo Scientific
OR2761-1G
OR2761-250MG
APOSOR2761-1GEA
6220
CZK
APOSOR2761-1G
APOSOR2761-250MG
3,4-Dichloro-2,6-dinitrophenol
3,4-Dichloro-2,6-dinitrophenol
Vzorec:
C₆H₂Cl₂N₂O₅ Bod tání: 168 °C Storage Temperature: Ambient |
MDL Number:
MFCD09836174 CAS číslo: 1664-10-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...