2-(Chloromethyl)-5-(4-chlorophenyl)-1,3,4-oxadiazole
Dodavatel: Apollo Scientific
OR27333-1G
OR27333-250MG
OR27333-500MG
APOSOR27333-1GEA
8930
CZK
APOSOR27333-1G
APOSOR27333-250MG
APOSOR27333-500MG
2-(Chloromethyl)-5-(4-chlorophenyl)-1,3,4-oxadiazole
2-(Chloromethyl)-5-(4-chlorophenyl)-1,3,4-oxadiazole
Vzorec:
C₉H₆Cl₂N₂O Bod tání: 83…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00714902 CAS číslo: 24068-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...