Tisk…

4-Chloro-N-2-dihydroxybenzamide

Dodavatel: Apollo Scientific

OR27157-1G OR27157-250MG OR27157-500MG OR27157-2.5G OR27157-5G
APOSOR27157-1GEA 9330 CZK
APOSOR27157-1G APOSOR27157-250MG APOSOR27157-500MG APOSOR27157-2.5G APOSOR27157-5G
4-Chloro-N-2-dihydroxybenzamide
4-Chloro-N-2-dihydroxybenzamide
Vzorec: C₇H₆ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00178744
CAS číslo: 61799-78-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR