Tisk…

3-Cyano-2-mercapto-6-methylisonicotinic acid ethyl ester

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 3-cyano-6-methyl-2-sulphanylisonicotinate

OR27079-5G OR27079-1G
APOSOR27079-5GEA 9820 CZK
APOSOR27079-5G APOSOR27079-1G
3-Cyano-2-mercapto-6-methylisonicotinic acid ethyl ester
3-Cyano-2-mercapto-6-methylisonicotinic acid ethyl ester
Vzorec: C₁₀H₁₀N₂O₂S
MW: 222,27 g/mol
Bod tání: 213…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00052275
CAS číslo: 56891-69-1
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR