Tisk…

4'-Bromo-2-chloroacetanilide

Dodavatel: Apollo Scientific

OR26967-10G OR26967-5G
APOSOR26967-10GEA 5290 CZK
APOSOR26967-10G APOSOR26967-5G
4'-Bromo-2-chloroacetanilide
4'-Bromo-2-chloroacetanilide
Vzorec: C₈H₇BrClNO
Storage Temperature: Ambient
MDL Number: MFCD00018903
CAS číslo: 2564-02-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR