4-Bromo-2-methylbenzenesulphonyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Bromo-2-methylbenzene-1-sulphonyl chloride
OR26859-1G
OR26859-25G
OR26859-5G
APOSOR26859-1GEA
1780
CZK
APOSOR26859-1G
APOSOR26859-25G
APOSOR26859-5G
4-Bromo-2-methylbenzenesulphonyl chloride 97%
4-Bromo-2-methylbenzenesulphonyl chloride
Vzorec:
C₇H₆BrClO₂S Bod varu: 164 °C/15mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00219927 CAS číslo: 139937-37-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...