Tisk…

2,6-Dibromo-4-chloroaniline 97%

Dodavatel: Apollo Scientific

Synonyms: 4-Chloro-2,6-dibromoaniline, 1-Amino-4-chloro-2,6-dibromobenzene

OR26854-100G OR26854-25G
APOSOR26854-100GEA 2570 CZK
APOSOR26854-100G APOSOR26854-25G
2,6-Dibromo-4-chloroaniline 97%
2,6-Dibromo-4-chloroaniline
Vzorec: C₆H₄Br₂ClN
MW: 285,37 g/mol
Bod tání: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00051750
CAS číslo: 874-17-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR