Tisk…

4-Methyl-2,1,3-benzothiadiazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Methylbenzo[c][1,2,5]thiadiazole

OR26818-1G OR26818-5G
APOSOR26818-1GEA 2360 CZK
APOSOR26818-1G APOSOR26818-5G
4-Methyl-2,1,3-benzothiadiazole
4-Methyl-2,1,3-benzothiadiazole
Vzorec: C₇H₆N₂S
MW: 150,2009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00126471
CAS číslo: 1457-92-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR