Methyl-1H-1,2,3-benzotriazole-5-carboxylate 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR26811-25G
OR26811-5G
APOSOR26811-25GEA
13030
CZK
APOSOR26811-25G
APOSOR26811-5G
Methyl-1H-1,2,3-benzotriazole-5-carboxylate 98%
Methyl-1H-1,2,3-benzotriazole-5-carboxylate
Vzorec:
C₈H₇N₃O₂ MW: 177,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00174276 CAS číslo: 113053-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...