Boc-L-cyclopropylglycine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
N-(tert-Butoxycarbonyl)-L-cyclopropylglycine, Boc-cyclopropyl-Gly-OH, (S)-2-[(tert-Butoxycarbonyl)amino]-2-cyclopropylacetic acid, N-Boc-L-cyclopropylglycine, (S)-(Boc-amino)-cyclopropylacetic acid, (S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopropylacetic acid, N-Boc-2-cyclopropyl-L-glycine, (S)-2-(Boc-amino)-2-cyclopropylacetic acid
OR2677-1G
OR2677-5G
OR2677-10G
APOSOR2677-1GEA
2200
CZK
APOSOR2677-1G
APOSOR2677-5G
APOSOR2677-10G
Boc-L-cyclopropylglycine
Boc-L-cyclopropylglycine
Vzorec:
C₁₀H₁₇NO₄ Bod tání: 79 °C Bod vzplanutí: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659132 CAS číslo: 155976-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...