Tisk…

2-Methyl-3-pyridineboronic acid pinacol ester 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Methylpyridine-3-boronic acid pinacol ester, 2-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

OR2664-1G OR2664-5G
APOSOR2664-1GEA 1820 CZK
APOSOR2664-1G APOSOR2664-5G
2-Methyl-3-pyridineboronic acid pinacol ester 95%
2-Methyl-3-pyridineboronic acid pinacol ester
Vzorec: C₁₂H₁₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD08669594
CAS číslo: 1012084-56-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR