3-Bromo-4-hydroxybenzyl alcohol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-Bromo-4-hydroxyphenyl)methanol, 2-Bromo-4-(hydroxymethyl)phenol
OR2644-1G
OR2644-5G
APOSOR2644-1GEA
705
CZK
APOSOR2644-1G
APOSOR2644-5G
3-Bromo-4-hydroxybenzyl alcohol
3-Bromo-4-hydroxybenzyl alcohol
Vzorec:
C₇H₇BrO₂ MW: 203,04 g/mol Bod tání: 128…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016869 CAS číslo: 29922-56-3 EINECS: 249-957-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...