3-Methoxy-5-methylbenzonitrile
Dodavatel: Apollo Scientific
OR2637-1G
OR2637-250MG
OR2637-500MG
OR2637-2.5G
APOSOR2637-1GEA
9330
CZK
APOSOR2637-1G
APOSOR2637-250MG
APOSOR2637-500MG
APOSOR2637-2.5G
3-Methoxy-5-methylbenzonitrile
3-Methoxy-5-methylbenzonitrile
Vzorec:
C₉H₉NO Storage Temperature: Ambient |
MDL Number:
MFCD09998751 CAS číslo: 473923-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...