Tisk…

Cyclooctane-1,5-dione

Dodavatel: Apollo Scientific
Varování

OR26280-1G OR26280-250MG
APOSOR26280-1GEA 20630 CZK
APOSOR26280-1G APOSOR26280-250MG
Cyclooctane-1,5-dione
Cyclooctane-1,5-dione
Vzorec: C₈H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00101719
CAS číslo: 1489-74-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR