Tisk…

1-Oxaspiro[2.5]octane-2-carbonitrile 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR26258-5G OR26258-10G
APOSOR26258-5GEA 3310 CZK
APOSOR26258-5G APOSOR26258-10G
1-Oxaspiro[2.5]octane-2-carbonitrile 95%
1-Oxaspiro[2.5]octane-2-carbonitrile
Vzorec: C₈H₁₁NO
Bod varu: 84…85 °C/5mmHg
Hustota: 1,0275 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00101211
CAS číslo: 36929-66-5
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR