Tisk…

(4-Methyl-1,3-thiazol-2-yl)methanol 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Methylthiazol-2-yl)methanol

OR2605-250MG OR2605-1G
APOSOR2605-250MGEA 787 CZK
APOSOR2605-250MG APOSOR2605-1G
(4-Methyl-1,3-thiazol-2-yl)methanol 98%
(4-Methyl-1,3-thiazol-2-yl)methanol
Vzorec: C₅H₇NOS
Storage Temperature: Refrigerator
MDL Number: MFCD08236826
CAS číslo: 13750-63-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR