Tisk…

ω-Bromo-4-phenylacetophenone 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromo-4'-phenylacetophenone, Bromomethyl p-phenylphenyl ketone, α-Bromo-4-phenylacetophenone, Biphenyl-4-yl bromomethyl ketone, 4-Phenylphenacyl bromide

OR26026-100G OR26026-25G
APOSOR26026-100GEA 0 CZK
APOSOR26026-100G APOSOR26026-25G
ω-Bromo-4-phenylacetophenone 97%
ω-Bromo-4-phenylacetophenone
Vzorec: C₁₄H₁₁BrO
MW: 275,14 g/mol
Bod tání: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00000202
CAS číslo: 135-73-9
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR