Tisk…

2,5-Dimethylbenzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,5-Dimethylbenzene-1-sulphonyl chloride, p-Xylene-2-sulphonyl chloride

OR26000-25G OR26000-5G OR26000-100G
APOSOR26000-25GEA 1910 CZK
APOSOR26000-25G APOSOR26000-5G APOSOR26000-100G
2,5-Dimethylbenzenesulphonyl chloride
2,5-Dimethylbenzenesulphonyl chloride
Vzorec: C₈H₉ClO₂S
Bod varu: 120…125 °C
Bod tání: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00024875
CAS číslo: 19040-62-1
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR