2-Bromo-4-methylthiazole
Dodavatel: Apollo Scientific
|
Varování
|
OR2600-1G
OR2600-5G
APOSOR2600-1GEA
622
CZK
APOSOR2600-1G
APOSOR2600-5G
2-Bromo-4-methylthiazole
2-Bromo-4-methylthiazole
Vzorec:
C₄H₄BrNS MW: 178,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660130 CAS číslo: 7238-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...