Tisk…

Methyl-3-chlorosulphonyl-4-(isopropylsulphonyl)thiophene-2-carboxylate

Dodavatel: Apollo Scientific

OR25983-10G OR25983-1G OR25983-5G
APOSOR25983-10GEA 19680 CZK
APOSOR25983-10G APOSOR25983-1G APOSOR25983-5G
Methyl-3-chlorosulphonyl-4-(isopropylsulphonyl)thiophene-2-carboxylate
Methyl-3-chlorosulphonyl-4-(isopropylsulphonyl)thiophene-2-carboxylate
Vzorec: C₉H₁₁ClO₆S₃
Storage Temperature: Ambient
MDL Number: MFCD03407370
CAS číslo: 423768-47-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR