Tisk…

5-(4-Methoxyphenyl)-1,3-oxazole-4-carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-(4-Methoxyphenyl)oxazole-4-carboxylic acid

OR25969-250MG OR25969-100MG
APOSOR25969-250MGEA 1580 CZK
APOSOR25969-250MG APOSOR25969-100MG
5-(4-Methoxyphenyl)-1,3-oxazole-4-carboxylic acid
5-(4-Methoxyphenyl)-1,3-oxazole-4-carboxylic acid
Vzorec: C₁₁H₉NO₄
MW: 219,2 g/mol
Bod tání: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD03644147
CAS číslo: 89205-07-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR