Diphenylphosphinic acid 99%
Dodavatel: Apollo Scientific
|
Varování
|
OR25790-100G
OR25790-25G
OR25790-500G
APOSOR25790-100GEA
1160
CZK
APOSOR25790-100G
APOSOR25790-25G
APOSOR25790-500G
Diphenylphosphinic acid 99%
Diphenylphosphinic acid
Vzorec:
C₁₂H₁₁O₂P Bod tání: 193…195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002132 CAS číslo: 1707-03-5 EINECS: 216-948-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...